Learn More
R&D Systems™ Human AMIGO3 (NP_942015) VersaClone cDNA
Amphoterin-induced Gene and ORF 3
Brand: R&D Systems™ RDC1793
This item is not returnable.
View return policy
Description
The Human AMIGO3 (NP_942015) VersaClone cDNA clone contains the complete ORF for AMIGO3, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The AMIGO3 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.Order Info
Ships at ambient temperature
Specifications
cDNA | |
NP_942015 | |
cDNA is provided in 10mM Tris-Cl, pH 8.5 | |
386724 | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. | |
10 μg |
AMIGO3 | |
10 μg at 0.2 μg/μL | |
1528bp | |
adhesion molecule with Ig-like domain 3, ALI3, Alivin-3, AMIGO-3, amphoterin-induced gene and ORF 3, amphoterin-induced protein 3, KIAA1851, MGC120552 | |
Human |