missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Carboxypeptidase B2/CPB2 Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF6036
This item is not returnable.
View return policy
Description
Carboxypeptidase B2/CPB2 Polyclonal specifically detects Carboxypeptidase B2/CPB2 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Carboxypeptidase B2/CPB2 | |
Polyclonal | |
Unconjugated | |
Q96IY4 | |
CPB2 | |
Mouse myeloma cell line NS0-derived recombinant human Carboxypeptidase B2/CPB2 Phe23-Val423 (Ala169Thr) Accession # Q96IY4 | |
100 μg | |
Primary | |
Detects Carboxypeptidase B2/CPB2 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) CPA1, rhCPB1 and recombinant mouse CPB1 is observed. | |
Human | |
IgG |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
carboxypeptidase B2, carboxypeptidase B2 (plasma), carboxypeptidase B2 (plasma, carboxypeptidase U), Carboxypeptidase U, CPB2, CPU, CPUEC 3.4.17.20, EC 3.4.17, PCPB, Plasma carboxypeptidase B, TAFI, TAFIcarboxypeptidase B-like protein, Thrombin-activable fibrinolysis inhibitor, thrombin-activatable fibrinolysis inhibitor | |
Sheep | |
Affinity Purified | |
RUO | |
1361 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction