missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Clusterin Quantikine ELISA Kit
Human Clusterin Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Brand: R&D Systems™ DCLU00
This item is not returnable.
View return policy
Description
The Human Clusterin Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human Clusterin/APOJ in cell culture supernates (50 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul), saliva (25 ul), urine (13 ul).Specifications
3.12 - 200ng/mL | |
HRP | |
Solid Phase Sandwich ELISA | |
Natural and Recombinant Human Clusterin | |
Human | |
1191 | |
Cell Culture Supernates (50uL),Serum (10uL),EDTA Plasma (10uL),Heparin Plasma (10uL),Saliva (25uL),Urine (13uL) | |
40, 40, sulfated glycoprotein 2, Aging-associated gene 4 protein, aging-associated protein 4, apo-J, APOJ, Apolipoprotein J, CLI, CLIclusterin (complement lysis inhibitor, SP-40, CLU, clusterin, Complement cytolysis inhibitor, complement lysis inhibitor, Complement-associated protein SP-40, Ku70-binding protein 1, KUB1SGP2, MGC24903, NA1/NA2, SGP-2, SP-40, sulfated glycoprotein 2, Testosterone-repressed prostate message 2, testosterone-repressed prostate message 2, apolipoprotein J), TRPM-2, TRPM-2 TRPM2 |
1.05ng/mL | |
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules | |
Cell Culture Supernatant, Plasma (EDTA, Heparin), Saliva, Serum, Urine | |
Sufficient materials to run ELISAs on at least one 96-well plate | |
ELISA | |
1 Plate | |
Store the unopened product at 2° to 8°C. Do not use past expiration date. |