Learn More
Invitrogen™ Human Connexin 25 (aa 143-180) Control Fragment Recombinant Protein
Description
Highest antigen sequence indentity to the following orthologs: Mouse (68%), Rat (68%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-140048 (PA5-140048. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Specifications
Specifications
| Accession Number | Q6PEY0 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 375519 |
| Name | Human Connexin 25 (aa 143-180) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | bA136M9.1; connexin 25; connexin25; connexin-25; C x 25; Gap junction beta-7 protein; gap junction protein beta 7; gap junction protein, beta 7, 25 kDa; GJB7 |
| Common Name | Connexin 25 |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.