missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human CXCL13/BLC/BCA-1 Magnetic Luminex™ Performance Assay

Product Code. 30002114
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 30002114

Brand: R&D Systems™ LBAM801

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Product Type Magnetic Bead-based Multiplex Assay for the Luminex™ MAGPIX™ Platform
Quantity 1 Kit
Species Human
Binding Target CXCL13/BLC/BCA-1
Gene ID (Entrez) 10563
Content And Storage Store the unopened product at 2 - 8 °C. Do not use past expiration date.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.