missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human DKFZP586B1621 Full-length ORF (AAH01341, 1 a.a. - 575 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00026007-P01.25ug
This item is not returnable.
View return policy
Description
This gene is a member of the family of dihydroxyacetone kinases, which have a protein structure distinct from other kinases. The product of this gene phosphorylates dihydroxyacetone, and also catalyzes the formation of riboflavin 4',5'-phosphate (aka cyclin FMN) from FAD. Several alternatively spliced transcript variants have been identified, but the full-length nature of only one has been determined. [provided by RefSeq]
Sequence: MTSKKLVNSVAGCADDALAGLVACNPNLQLLQGHRVALRSDLDSLKGRVALLSGGGSGHEPAHAGFIGKGMLTGVIAGAVFTSPAVGSILAAIRAVAQAGTVGTLLIVKNYTGDRLNFGLAREQARAEGIPVEMVVIGDDSAFTVLKKAGRRGLCGTVLIHKVAGALAEAGVGLEEIAKQVNVVAKAMGTLGVSLSSCSVPGSKPTFELSADEVELGLGIHGEAGVRRIKMATADEIVKLMLDHMTNTTNASHVPVQPGSSVVMMVNNLGGLSFLELGIIADATVRSLEGRGVKIARALVGTFMSALEMPGISLTLLLVDEPLLKLIDAETTAAAWPNVAAVSITGRKRSRVAPAEPQEAPDSTAAGGSASKRMALVLERACSTLLGLEEHLNALDRAAGDGDCGTTRSRAARAIQEWLKEGPPPASPAQLLSKLSVLLLEKMGGSSGALYGLFLTAAAQPLKAKTSLPAWSAAMDAGLEAMQKYGKAAPGDRTMLDSLWAAGQELQAWKSPGADLLQVLTKAVKSAEAAAEATKNMEAGAGRASYISSARLEQPDPGAVAAAAILRAILEVLQSSpecifications
AAH01341 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
88.99kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp586B1621/MGC5621 | |
DAK | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
26007 | |
DKFZP586B1621 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MTSKKLVNSVAGCADDALAGLVACNPNLQLLQGHRVALRSDLDSLKGRVALLSGGGSGHEPAHAGFIGKGMLTGVIAGAVFTSPAVGSILAAIRAVAQAGTVGTLLIVKNYTGDRLNFGLAREQARAEGIPVEMVVIGDDSAFTVLKKAGRRGLCGTVLIHKVAGALAEAGVGLEEIAKQVNVVAKAMGTLGVSLSSCSVPGSKPTFELSADEVELGLGIHGEAGVRRIKMATADEIVKLMLDHMTNTTNASHVPVQPGSSVVMMVNNLGGLSFLELGIIADATVRSLEGRGVKIARALVGTFMSALEMPGISLTLLLVDEPLLKLIDAETTAAAWPNVAAVSITGRKRSRVAPAEPQEAPDSTAAGGSASKRMALVLERACSTLLGLEEHLNALDRAAGDGDCGTTRSRAARAIQEWLKEGPPPASPAQLLSKLSVLLLEKMGGSSGALYGLFLTAAAQPLKAKTSLPAWSAAMDAGLEAMQKYGKAAPGDRTMLDSLWAAGQELQAWKSPGADLLQVLTKAVKSAEAAAEATKNMEAGAGRASYISSARLEQPDPGAVAAAAILRAILEVLQS | |
RUO | |
DAK | |
Wheat Germ (in vitro) | |
GST | |
Liquid |