Learn More
Abnova™ Human FBXL6 Full-length ORF (NP_036294.1, 1 a.a. - 539 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00026233-P01.25ug
Description
This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbls class and, in addition to an F-box, contains several tandem leucine-rich repeats. Alternative splicing of this gene generates 2 transcript variants. [provided by RefSeq]
Sequence: MAAPASRQVRRRARAAPRPRSAEDWWWDRLAPRGSGYHLLQSDSMLLVLSEPGPARPRAQRRASRRTPRQPPRGPSAAAKPKAGLRSEAAAAPAPAPAPTPTPEEGPDAGWGDRIPLEILVQIFGLLVAADGPMPFLGRAARVCRRWQEAASQPALWHTVTLSSPLVGRPAKGGVKAEKKLLASLEWLMPNRFSQLQRLTLIHWKSQVHPVLKLVGECCPRLTFLKLSGCHGVTADALVMLAKACCQLHSLDLQHSMVESTAVVSFLEEAGSRMRKLWLTYSSQTTAILGALLGSCCPQLQVLEVSTGINRNSIPLQLPVEALQKGCPQLQVLRLLNLMWLPKPPGRGVAPGPGFPSLEELCLASSTCNFVSNEVLGRLLHGSPNLRLLDLRGCARITPAGLQDLPCRELEQLHLGLYGTSDRLTLAKEGSPFLTQKWCHTLRELDLSGQGFSEKDLEQALAAFLSTPGGSHPALCSLNLRGTRVTPSTVSSVISSCPGLLYLNLESCRCLPRGLKRAYRGLEEVQWCLEQLLTSPSPSSpecifications
NP_036294.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
85kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FBL6/FBL6A/PP14630 | |
FBXL6 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
26233 | |
FBXL6 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAAPASRQVRRRARAAPRPRSAEDWWWDRLAPRGSGYHLLQSDSMLLVLSEPGPARPRAQRRASRRTPRQPPRGPSAAAKPKAGLRSEAAAAPAPAPAPTPTPEEGPDAGWGDRIPLEILVQIFGLLVAADGPMPFLGRAARVCRRWQEAASQPALWHTVTLSSPLVGRPAKGGVKAEKKLLASLEWLMPNRFSQLQRLTLIHWKSQVHPVLKLVGECCPRLTFLKLSGCHGVTADALVMLAKACCQLHSLDLQHSMVESTAVVSFLEEAGSRMRKLWLTYSSQTTAILGALLGSCCPQLQVLEVSTGINRNSIPLQLPVEALQKGCPQLQVLRLLNLMWLPKPPGRGVAPGPGFPSLEELCLASSTCNFVSNEVLGRLLHGSPNLRLLDLRGCARITPAGLQDLPCRELEQLHLGLYGTSDRLTLAKEGSPFLTQKWCHTLRELDLSGQGFSEKDLEQALAAFLSTPGGSHPALCSLNLRGTRVTPSTVSSVISSCPGLLYLNLESCRCLPRGLKRAYRGLEEVQWCLEQLLTSPSPS | |
RUO | |
FBXL6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |