missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Galectin-3 Quantikine ELISA Kit
Human Galectin-3 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Brand: R&D Systems™ DGAL30
This item is not returnable.
View return policy
Description
The Human Galectin-3 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human Galectin-3 in cell culture supernates (50 ul), serum (50 ul), edta plasma (50 ul), heparin plasma (50 ul).
Specifications
0.31 to 10 ng/mL | |
HRP | |
Cell Culture Supernatant, Plasma (EDTA), Plasma (EDTA, Heparin), Plasma (Heparin), Serum, Serum | |
ELISA | |
LGALS3 | |
Cell Culture Supernates (50uL),Serum (50uL),EDTA Plasma (50uL),Heparin Plasma (50uL) | |
AGE-R3, Carbohydrate-binding protein 35, CBP 35,35 kDa lectin, CBP35, Gal-3, GAL3, galactin-3, Galactose-specific lectin 3, Galactoside-binding protein, GALBPCBP35, galectin-3, Galectin3, GALIG, IgE-binding protein, L29, L31, L-31, Laminin-binding protein, Lectin L-29, lectin, galactoside-binding, soluble, 3, LGALS2, LGALS3, Mac-2, Mac-2 antigen, MAC2GAL3 |
0.085 ng/mL | |
Solid Phase Sandwich ELISA | |
Natural and Recombinant Human Galectin-3. This assay cross-reacts 58% with Mouse Galectin-3. | |
3958 | |
1 Kit | |
Store the unopened product at 2°C to 8°C. Do not use past expiration date. | |
Galectin-3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
R&D Systems™ Human Galectin-3 Quantikine ELISA Kit > 1 Kit
Spot an opportunity for improvement?Share a Content Correction