Learn More
Invitrogen™ Human GALNT5 (aa 74-198) Control Fragment Recombinant Protein
Descrizione
Highest antigen sequence indentity to the following orthologs: Mouse (51%), Rat (51%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-52641 (PA5-52641. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Specifica
Specifica
| Accession Number | Q7Z7M9 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 11227 |
| Name | Human GALNT5 (aa 74-198) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | GALNACT5; GalNAc-T5; GALNT5; Polypeptide GalNAc transferase 5; polypeptide N-acetylgalactosaminyltransferase 5; pp-GaNTase 5; protein-UDP acetylgalactosaminyltransferase 5; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 5; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 5 (GalNAc-T5) |
| Common Name | GALNT5 |
| Vedi altri risultati |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.