missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human GJA8 Partial ORF (NP_005258, 301 a.a. - 410 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00002703-Q01.10ug
This item is not returnable.
View return policy
Description
Sequence: EEKISTGPLGDLSRGYQETLPSYAQVGAQEVEGEGPPAEEGAEPEVGEKKEEAERLTTEEQEKVAVPEGEKVETPGVDKEGEKEEPQSEKVSKQGLPAEKTPSLCPELTTSpecifications
NP_005258 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
EEKISTGPLGDLSRGYQETLPSYAQVGAQEVEGEGPPAEEGAEPEVGEKKEEAERLTTEEQEKVAVPEGEKVETPGVDKEGEKEEPQSEKVSKQGLPAEKTPSLCPELTT | |
RUO | |
GJA8 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
2703 | |
GJA8 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CAE/CAE1/CX50/CZP1/MP70 | |
GJA8 | |
Recombinant | |
wheat germ expression system |