missing translation for 'onlineSavingsMsg'
Learn More

Human Glypican 1 Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

221.00 € - 471.00 €

Specifications

Antigen Glypican 1
Clone 1019718
Dilution Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18732387
GREENER_CHOICE
View Documents
R&D Systems
MAB45192-SP
25 μg
221.00 €
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18792427
GREENER_CHOICE
View Documents
R&D Systems
MAB45192-100
100 μg
471.00 €
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Glypican 1 Monoclonal antibody specifically detects Glypican 1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry
Specifications

Specifications

Glypican 1
Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
FLJ38078, glypican, glypican 1, glypican proteoglycan 1, glypican-1, GPC1
Mouse myeloma cell line NS0­derived human Glypican 1, Asp24-Ser530, Accession # P35052
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
1019718
Flow Cytometry, Immunocytochemistry
Unconjugated
Mouse
Human
P35052
2817
IgG1
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.