missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human HBLD1 Full-length ORF (NP_919255.1, 1 a.a. - 154 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00122961-P01.10ug
This item is not returnable.
View return policy
Description
Sequence: MAAAWGSSLTAATQRAVTPWPRGRLLTASLGPQARREASSSSPEAGEGQICLTDSCVQRLLEITEGSEFLRLQVEGGGCSGFQYKFSLDTVINPDDRVFEQGGARVVVDSDSLAFVKGAQVDFSQELIRSSFQVLNNPQAQQGCSCGSSFSIKLSpecifications
NP_919255.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
42.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HBLD1/ISA2/c14_5557 | |
ISCA2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
122961 | |
HBLD1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAAAWGSSLTAATQRAVTPWPRGRLLTASLGPQARREASSSSPEAGEGQICLTDSCVQRLLEITEGSEFLRLQVEGGGCSGFQYKFSLDTVINPDDRVFEQGGARVVVDSDSLAFVKGAQVDFSQELIRSSFQVLNNPQAQQGCSCGSSFSIKL | |
RUO | |
ISCA2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |