missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human Heat Stable FGF-basic (FGF-2/bFGF) (154 aa) Recombinant Protein, PeproTech®

Product Code. 17867058
Click to view available options
Quantity:
1 mg
10 μg
100 μg
250 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
250µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 17867058

Brand: Gibco™ 10018BHS500UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

FGF-basic loses its activity over time in solution at physiological temperatures and needs to be re-supplemented periodically to maintain its performance. As a result of selected amino acid substitutions, Heat Stable FGF-basic retains full biological activity and also maintains full activity over extended periods of time in solution at 370 C. These properties make Heat Stable FGF-basic a superior choice for use in cell culture media applications. Recombinant Heat Stable FGF-basic is a 17.2 kDa protein containing 154 amino acids. Human FGF-basic (FGF-2/bFGF) (154 aa), Recombinant Protein PeproTech® was developed under a license from Enantis. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

FGF2 (FGFb, fibroblast growth factor basic) belongs to the fibroblast growth factor (FGF) family, and interacts with high-affinity transmembrane receptors to influence cell proliferation and tissue neovascularization. FGF2 exists as five isoforms with distinct intracellular localizations and functions. The 18 kDa isoform is predominantly cytosolic and acts through cell surface receptors, whereas the 22, 22.5, 24 and 34 kDa isoforms are nuclear and may signal independent of transmembrane receptor pathways. In humans, the gene is located on the q arm of chromosome 4. FGF2 has been implicated in diverse biological processes, such as limb and nervous system development, wound healing, and tumor growth. The mRNA for FGF2 contains multiple polyadenylation sites, and is alternatively translated from non-AUG and AUG initiation codons, resulting in five different isoforms with distinct properties. The CUG-initiated isoforms are localized in the nucleus and are responsible for the intracrine effect, whereas, the AUG-initiated form is mostly cytosolic and is responsible for the paracrine and autocrine effects of this FGF. Diseases associated with FGF2 dysfunction include Kaposi Sarcoma and corneal neovascularization.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P09038
For Use With (Application) Functional Assay
Format Lyophilized
Formulation protein with no preservative
Gene ID (Entrez) 2247
Molecular Weight (g/mol) 17.2 kDa
Name Human Heat Stable FGF-basic (FGF-2/bFGF) (154 aa)
Purification Method HPLC, SDS-PAGE
Quantity 500 μg
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias Basic fibroblast growth factor; basic fibroblast growth factor bFGF; bFGF; FGF; fgf basic; Fgf2; Fgf-2; Fgfb; FGF-b; Fibroblast growth factor; fibroblast growth factor 2; fibroblast growth factor 2 (basic); HBGF-2; Heparin-binding growth factor 2; H-FGF-b-147; H-FGF-b-154; M-FGF-b; Prostatic growth factor; prostatropin
Common Name FGF2
Gene Symbol FGF2
Biological Activity Determined by a cell proliferation assay using Balb/c 3t3 cells. The expected ED50 range is 0.05-0.25 ng/ml
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence Thermostable variant of human FGF-basic (154 amino acids) with proprietary amino acid substitutions.
Content And Storage -20°C
Activity Determined by a cell proliferation assay using Balb/c 3t3 cells. The expected ED50 range is 0.05-0.25 ng/ml
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.