missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human IL-4 Recombinant Protein
Recombinant Human Interleukin 4 (IL4) is a bioactive protein intended for use in cell culture applications
287.00 € - 6520.00 €
Specifications
For Use With (Application) | Cell Culture |
---|---|
Gene ID (Entrez) | 3565 |
Molecular Weight (g/mol) | 14.9 kDa |
Purification Method | Sequential Chromatography |
Gene Alias | IL-4 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10474553
|
Gibco™
PHC0044 |
10μg |
287.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10593765
|
Gibco™
PHC0045 |
25μg |
550.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10069352
|
Gibco™
PHC0041 |
100μg |
1954.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10474363
|
Gibco™
PHC0043 |
1mg |
6520.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Human IL-4 Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
IL4 promotes the proliferation and differentiation of activated B cells. In addition, IL4 up-regulates class II MHC antigen expression and IgE receptors.- High purity: no interference from other proteins or contaminants
- High biological activity: more results with less protein
- Proven compatibility: Gibco proteins bioassayed with Gibco media
- High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
- High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by measuring the dose-dependent proliferation of human TF-1 cells
- Proven Compatibility: Gibco proteins are bioassayed with Gibco media
Specifications
Cell Culture | |
14.9 kDa | |
IL-4 | |
Store in refrigerator (2–8°C). | |
< 0.1 ng/μg | |
E. coli | |
Recombinant, Ligand | |
Interleukins | |
Gibco™ |
3565 | |
Sequential Chromatography | |
Human | |
Carrier-Free | |
Wet Ice | |
Cytokines & Receptors | |
Lyophilized | |
Immunology, Stem Cell Research, Inflammation | |
95 % |
For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.