missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human IL-7 Recombinant Protein, PeproTech®
Click to view available options
Quantity:
10 μg
100 μg
2 μg
2 x 500 μg
250 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
250µg
2µg
500µg
50µg
Pack of 2
Description
200-07-1MG will be provided as 2 x 500 μg (200-07-500UG). Recombinant Human IL-7 is a 17.5 kDa protein containing 153 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Interleukin 7 (IL-7) is a hematopoietic cytokine that is constitutively produced by stromal cells from the bone marrow and thymus. IL-7 signals through the IL-7R to promote the differentiation of hematopoietic stem cells to lymphoid precursor cells, which give rise to T cells, B cells, and natural killer cells.
Specifications
Specifications
| Accession Number | P13232, P16871 |
| For Use With (Application) | Control, Western Blot Control, ELISA Standard, Bioactivity |
| Formulation | Protein with no preservative |
| Gene ID (Entrez) | 3574, 3575 |
| Molecular Weight (g/mol) | 17.5 kDa |
| Name | Human IL-7 |
| Purification Method | Purified |
| Quantity | 250 μg |
| Source | E. coli |
| Storage Requirements | -20°C |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction