missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human IL17A (NP_002181.1, 24 a.a. - 155 a.a.) Partial Recombinant Protein
Used for Dot, ELISA, IHC, SDS-PAGE, WB
Brand: Abnova™ P3814.10ug
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a proinflammatory cytokine produced by activated T cells. This cytokine regulates the activities of NF-kappaB and mitogen-activated protein kinases. This cytokine can stimulate the expression of IL6 and cyclooxygenase-2 (PTGS2/COX-2), as well as enhance the production of nitric oxide (NO). High levels of this cytokine are associated with several chronic inflammatory diseases including rheumatoid arthritis, psoriasis and multiple sclerosis. [provided by RefSeq]
Specifications
NP_002181.1 | |
Liquid | |
IL17A (Human) Recombinant Protein | |
10% SDS-PAGE Result | |
Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CTLA8/IL-17/IL-17A/IL17 | |
IL17A | |
Recombinant | |
Escherichia coli expression system | |
>95% by SDS-PAGE |
Dot Blot, ELISA, Immunohistochemistry, SDS-PAGE, Western Blot | |
3605 | |
Escherichia coli expression system | |
10 μg | |
RUO | |
IL17A | |
E. coli | |
None | |
Liquid |