Learn More
Human L1CAM Alexa Fluor™ 750-conjugated Antibody, R&D Systems™
Rabbit Monoclonal Antibody
Brand:  R&D Systems FAB7773S-100UG
Product Code. 18768566
  • 521.00 € / 100µg
Please to purchase this item. Need a web account? Register with us today!
Explore more special offers
This item is not returnable. View return policy
Description
Description
L1CAM Monoclonal antibody specifically detects L1CAM in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
L1CAM
Monoclonal
Alexa Fluor 750
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
antigen identified by monoclonal R1, CAML1, CAML1N-CAML1, CD171, CD171 antigen, HSAS, HSAS1, L1 cell adhesion molecule, MASA, MIC5, N-CAM-L1, NCAM-L1, neural cell adhesion molecule L1, S10, SPG1
Mouse myeloma cell line NS0­ derived Human L1CAM, (Ile20­Glu1120) & (Arg864­Glu1120), Accession # CAA42508
100 μg
Primary
Human
Purified
Flow Cytometry
2702C
Flow Cytometry
CAA42508
Rabbit
Protein A or G purified from cell culture supernatant
RUO
3897
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Human L1CAM Alexa Fluor™ 750-conjugated Antibody, R&D Systems™ > 100 μg; Alexa Fluor 750