missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MCP-1 ELISA Development Kit (TMB), PeproTech®

Product Code. 30089324
Click to view available options
Quantity:
10 x 96 Tests
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 30089324

Brand: Invitrogen™ 900T31K

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

This gene is one of several cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The protein encoded by this gene is structurally related to the CXC subfamily of cytokines. Members of this subfamily are characterized by two cysteines separated by a single amino acid. This cytokine displays chemotactic activity for monocytes and basophils but not for neutrophils or eosinophils. It has been implicated in the pathogenesis of diseases characterized by monocytic infiltrates, like psoriasis, rheumatoid arthritis and atherosclerosis. It binds to chemokine receptors CCR2 and CCR4.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P13500
Assay Range Lot-specific
Assay Sensitivity Lot-specific
Conjugate HRP
Product Type ELISA
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 6347
Gene Symbol Ccl2
Kit Contents Capture Antibody, Detection Antibody, Protein Standard, HRP-conjugate
Label or Dye HRP
Quantity 10 x 96 Tests
Regulatory Status RUO
Storage Requirements Refer to product documentation for component specific storage temperature.
Target CCL2 (MCP-1)
Test Time 1 hrs 45 mins
Total Assay Time 24 hrs 30 mins
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.