missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human MEI1 Full-length ORF (AAH32248.1, 1 a.a. - 238 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 € - 508.00 €
Specifications
Accession Number | AAH32248.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 150365 |
Molecular Weight (g/mol) | 52kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16128613
|
Abnova™
H00150365-P01.10ug |
10 ug |
335.00 €
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16138613
|
Abnova™
H00150365-P01.25ug |
25 ug |
508.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence: MLALTLAKADSPRTALLCSAWLLTASFSAQQHKGSLQVHQTLSVEMDQVLKALSFPKKKAALLSAAILCFLRTALRQSFSSALVALVPSGAQPLPATKDTVLAPLRMSQVRSLVIGLQNLLVQKDPLLSQACVGCLEALLDYLDARSPDIALHVASQPWNRFLLFTLLDAGENSFLRPEILRLMTLLQSMGHLADHSMAQTLQASLEGLPPSTSSGQPPLQDMLCLGGVAVSLSHIRNSpecifications
AAH32248.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
52kDa | |
Glutathione Sepharose 4 Fast Flow | |
MLALTLAKADSPRTALLCSAWLLTASFSAQQHKGSLQVHQTLSVEMDQVLKALSFPKKKAALLSAAILCFLRTALRQSFSSALVALVPSGAQPLPATKDTVLAPLRMSQVRSLVIGLQNLLVQKDPLLSQACVGCLEALLDYLDARSPDIALHVASQPWNRFLLFTLLDAGENSFLRPEILRLMTLLQSMGHLADHSMAQTLQASLEGLPPSTSSGQPPLQDMLCLGGVAVSLSHIRN | |
RUO | |
MEI1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
150365 | |
MEI1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC40042 | |
MEI1 | |
Recombinant | |
wheat germ expression system |