missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human NAP1L3 Full-length ORF (AAH34954.1, 1 a.a. - 506 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00004675-P01.25ug
This item is not returnable.
View return policy
Description
This gene encodes a member of the nucleosome assembly protein (NAP) family. It is linked closely to a region of genes responsible for several X-linked mental retardation syndromes. Alternatively spliced transcript variants have been found for this gene, but the biological validity of the variants has not been determined. [provided by RefSeq]
Sequence: MAEADFKMVSEPVAHGVAEEEMASSTSDSGEESDSSSSSSSTSDSSSSSSTSGSSSGSGSSSSSSGSTSSRSRLYRKKRVPEPSRRARRAPLGTNFVDRLPQAVRNRVQALRNIQDECDKVDTLFLKAIHDLERKYAELNKPLYDRRFQIINAEYEPTEEECEWNSEDEEFSSDEEVQDNTPSEMPPLEGEEEENPKENPEVKAEEKEVPKEIPEVKDEEKEVAKEITEVKAEEKADSKDCMEATPEVKEDPKEVPQVKADDKEQPKATEAKARAAVRETHKRVPEERLQDSVDLKRARKGKPKREDPKGIPDYWLIVLKNVDKLGPMIQKYDEPILKFLSDVSLKFSKPGQPVSYTFEFHFLPNPYFRNEVLVKTYIIKAKPDHNDPFFSWGWEIEDCKGCKIDWRRGKDVTVTTTQSRTTATGEIEIQPRVVPNASFFNFFSPPEIPMIGKLEPREDAILDEDFEIGQILHDNVILKSIYYYTGEVNGTYYQFGKHYGNKKYRKSpecifications
AAH34954.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
84kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MB20/MGC26312/NPL3 | |
NAP1L3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
4675 | |
NAP1L3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAEADFKMVSEPVAHGVAEEEMASSTSDSGEESDSSSSSSSTSDSSSSSSTSGSSSGSGSSSSSSGSTSSRSRLYRKKRVPEPSRRARRAPLGTNFVDRLPQAVRNRVQALRNIQDECDKVDTLFLKAIHDLERKYAELNKPLYDRRFQIINAEYEPTEEECEWNSEDEEFSSDEEVQDNTPSEMPPLEGEEEENPKENPEVKAEEKEVPKEIPEVKDEEKEVAKEITEVKAEEKADSKDCMEATPEVKEDPKEVPQVKADDKEQPKATEAKARAAVRETHKRVPEERLQDSVDLKRARKGKPKREDPKGIPDYWLIVLKNVDKLGPMIQKYDEPILKFLSDVSLKFSKPGQPVSYTFEFHFLPNPYFRNEVLVKTYIIKAKPDHNDPFFSWGWEIEDCKGCKIDWRRGKDVTVTTTQSRTTATGEIEIQPRVVPNASFFNFFSPPEIPMIGKLEPREDAILDEDFEIGQILHDNVILKSIYYYTGEVNGTYYQFGKHYGNKKYRK | |
RUO | |
NAP1L3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |