missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human NEDD9 Partial ORF (NP_006394, 101 a.a. - 200 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00004739-Q01.25ug
This item is not returnable.
View return policy
Description
Sequence: PRDTIYQVPPSYQNQGIYQVPTGHGTQEQEVYQVPPSVQRSIGGTSGPHVGKKVITPVRTGHGYVYEYPSRYQKDVYDIPPSHTTQGVYDIPPSSAKGPVSpecifications
NP_006394 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
PRDTIYQVPPSYQNQGIYQVPTGHGTQEQEVYQVPPSVQRSIGGTSGPHVGKKVITPVRTGHGYVYEYPSRYQKDVYDIPPSHTTQGVYDIPPSSAKGPV | |
RUO | |
NEDD9 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
4739 | |
NEDD9 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CAS-L/CAS2/CASL/CASS2/HEF1/dJ49G10.2/dJ761I2.1 | |
NEDD9 | |
Recombinant | |
wheat germ expression system |