Learn More
Gibco™ Human Noggin Recombinant Protein, PeproTech®
Beschreibung
120-10C-1MG will be provided as 2 x 500 μg (120-10C-500UG). Recombinant Human Noggin is a 46 kDa disulfide-linked homodimer consisting of two 205 amino acid polypeptide chains. Monomeric glycosylated noggin migrates at an apparent molecular weight of approximately 28.0-33.0 kDa by SDS PAGE analysis under reducing conditions. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Spezifikation
Spezifikation
| Accession Number | Q13253 |
| For Use With (Application) | Functional Assay |
| Formulation | protein with no preservative |
| Gene ID (Entrez) | 9241 |
| Molecular Weight (g/mol) | 46 |
| Name | Human Noggin |
| Quantity | 2 x 500 μg |
| Source | HEK293 cells |
| Storage Requirements | -20°C |
| Regulatory Status | RUO |
| Mehr anzeigen |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.