Learn More
R&D Systems™ Human OAT1 (NP_004781) VersaClone cDNA
Sequence verified insert equipped with translated sequences for the insert and entire plasmid DNA.
Brand: R&D Systems™ RDC2587
This item is not returnable.
View return policy
Description
- The Human OAT1 (NP_004781) VersaClone cDNA clone contains the complete ORF for SLC22A6, along with a Kozak consensus sequence for optimal translation initiation
- The gene insert is flanked with convenient multiple cloning sites, which can be used to easily cut and transfer the gene cassette into your desired expression vector
- The SLC22A6 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided
Specifications
hOAT1 | |
10 μg at 0.2 μg/μL | |
9356 | |
Human |
NP_004781 | |
cDNA is provided in 10 mM Tris-Cl, pH 8.5 | |
Store the unopened product at -20 to -80°C | |
10 μg |