missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human RANBP2 (aa 1-18) Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP137
This item is not returnable.
View return policy
Description
Human RanBP2 (reaidues 1-18) Synthetic Peptide, PEP-137, from Invitrogen.
PEP-137 is an 18 amino acid synthetic peptide whose sequences are derived from human RanBP2 protein. The sequence of this peptide is (amino to carboxy terminus): M (1)-R-R-S-K-A-D-V-E-R-Y-I-A-S-V-Q-G-S (18)-CThis peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and human RanBP2, catalog # PA1-082. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∼70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 ml of distilled water.Specifications
5903 | |
purified | |
Human RANBP2 (aa 1-18) | |
-20°C. Avoid freeze-thaw cycles. | |
Lyophilized | |
Control | |
Yes | |
Peptide | |
RUO - research use only |
Human | |
P49792 | |
0.5 mg/mL | |
Synthetic Peptide | |
peptide with no preservative | |
RANBP2 | |
No preservative | |
50 μg | |
M(1)-R-R-S-K-A-D-V-E-R-Y-I-A-S-V-Q-G-S(18)-C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction