missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RNF19 Partial ORF (NP_056250, 739 a.a. - 837 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00025897-Q01.10ug

 View more versions of this product

Product Code. 16137872

  • 335.00 € / 10µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The protein encoded this gene contains two RING-finger motifs and an IBR (in between RING fingers) motif. This protein is an E3 ubiquintin ligase that is localized in Lewy bodies (LBs), a characteristic neuronal inclusion in Parkinson's disease (PD) brains. This protein interacts with UBE2L3/UBCH7 and UBE2E2/UBCH8, but not other ubiquitin-conjugating enzymes. This protein is found to bind and ubiquitylate synphilin 1 (SNCAIP), which is a interacting protein of alpha synuclein in neurons, and a major component of LB. Alternatively spliced transcript variants encoding the same protein have been reported. [provided by RefSeq]

Sequence: MKTSCSHGSSDYHTRFATVNILPEVENDRLENSPHQCSISVVTQTASCSEVSQLNHIAEEHGNNGIKPNVDLYFGDALKETNNNHSHQTMELKVAIQTE
Specifications

Specifications

NP_056250
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MKTSCSHGSSDYHTRFATVNILPEVENDRLENSPHQCSISVVTQTASCSEVSQLNHIAEEHGNNGIKPNVDLYFGDALKETNNNHSHQTMELKVAIQTE
RUO
RNF19A
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
25897
RNF19 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp566B1346/DORFIN/RNF19
RNF19A
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions