missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech™ Human Total MMP-3 ELISA Kit
KE00160 is a solid phase sandwich Enzyme Linked-Immuno-Sorbent Assay (Sandwich ELISA). The Total MMP3 ELISA kit is to be used to detect and quantify protein levels of endogenous Total MMP3.
Brand: Proteintech™ KE00160-96T
This item is not returnable.
View return policy
Description
- MMP-3 (matrix metallopeptidase 3), also named as Stromelysin 1 or STR1, is a member of matrix metalloproteinase (MMP) family.
- The MMP family of enzymes is comprised of critically important extracellular matrix remodeling proteases whose activity has been implicated in normal embryogenesis and tissue remodelling and in many diseases such as arthritis, cancer, periodontitis, glomerulonephritis, encephalomyelitis, atherosclerosis and tissue ulceration.
- These proteases have come to represent important therapeutic and diagnostic targets for the treatment and detection of human cancers. MMP-3 is secreted from the cells as a proenzyme. The proenzyme has been shown to stimulate plasminogen activation.
- The active MMP-3 is capable of cleaving types III, IV, IX and X collagen, aggrecan, fibronectin, laminin, IGFBP-3, serpins, and IL-1β.
- The assay recognizes human Total MMP3. An antibody specific for Total MMP3 has been pre-coated onto the microwells. The Total MMP3 protein in samples is captured by the coated antibody after incubation.
- Following extensive washing, another antibody specific for Total MMP3 is added to detect the captured Total MMP3 protein.
- For signal development, Sterptravidin-HRP is added, followed by Tetramethyl-benzidine (TMB) reagent.
- Solution containing sulfuric acid is used to stop color development and the color intensity which is proportional to the quantity of bound protein is measurable at 450nm with the correction wavelength set at 630 nm.
Specifications
BC069676 | |
18.4 pg/mL | |
Sandwich ELISA, Double Antibody | |
Plasma, Serum | |
Human | |
Human | |
<10% | |
Blue ice | |
4.5 hr. |
31.25 to 2000 pg/mL | |
ISO 13485 | |
Coated ELISA Kit | |
DAB | |
4314 | |
Solid | |
<10% | |
-20°C to +4°C | |
P08254 |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- 96TEST Human Total MMP-3 ELISA Kit
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 1
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
Precautionary Statement
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P234-Keep only in original container.
- P264-Wash skin thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P330-Rinse mouth.
- P362-Take off contaminated clothing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Sulphuric Acid, Tetramethylbenzidine
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction