Learn More
R&D Systems™ Human VEGF High Sensitivity Magnetic Luminex™ Performance Assay
Luminex bead-based multiplex assays are designed to provide accurate, reproducible results for every target analyte.
Brand: R&D Systems™ LHSCM293
Description
- Magnetic format allows for easier wash steps - Undergoes similar validation testing as Quantikine™ ELISA Kits - Precise and Specific - Requires a small sample volume (<50μL) - Run your assay in just 3-5 hours - Mass-calibrated standards for consistent results with every new lot of material - Enough reagents for one 96-well plate
Specifications
Magnetic Bead-based Multiplex Assay for the Luminex™ MAGPIX™ Platform | |
Human | |
Magnetic | |
2.20 - 2300pg/mL | |
VEGFA | |
Serum (50μL),EDTA Plasma (50μL),Heparin Plasma (50μL) | |
Store the unopened product at 2° to 8°C. Do not use past expiration date. |
1 Kit | |
VEGF | |
1.35pg/mL | |
7422 | |
Serum, Plasma (EDTA, Heparin) | |
|
Your input is important to us. Please complete this form to provide feedback related to the content on this product.