Learn More
Invitrogen™ Human ZMYM1 Synthetic Peptide
Product Code. 15205209
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
For Research Use Only. All usage must comply with product instructions.
Product Code. 15205209
Brand: Invitrogen™ PEP1203
Please to purchase this item. Need a web account? Register with us today!
For Research Use Only. All usage must comply with product instructions.
Synthetic Peptide
ZMYM1 Synthetic Peptide, PEP-1203, from Invitrogen.
Zinc-finger proteins contain DNA-binding domains characterized by the unique role of zinc and have a wide variety of functions such as transcriptional activation or repression. The protein folding and the DNA binding ability are governed by the coordination of a zinc ion. As a member of the MYM (myeloproliferative and mental retardation) gene family, ZMYM1 is widely expressed in different tissues in eukaryotes under several forms derived by alternative splicing. While its function remains unknown, the related protein ZMYM2 has been shown to associate with and stabilize the LSD1-CoREST-HDAC1 (LCH) complex of chromatin through its MYM-type zinc fingers, thereby enhancing the transcriptional repression of several genes, suggesting that ZMYM1 may play a similar role.
TRUSTED_SUSTAINABILITY
Specifications
Gene ID (Entrez) 79830
Species Human
Purification Method purified
Accession Number Q5SVZ6
Common Name Human ZMYM1
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Symbol ZMYM1
Lyophilized No
Preservative 0.02% sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Storage Buffer PBS