missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ZNF21 Partial ORF (NP_008893, 81 a.a. - 190 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00007569-Q01.25ug
This item is not returnable.
View return policy
Description
Sequence: VEECPAEGKIPFWNFPEVCQVDEQIERQHQDDQDKCLLMQVGFSDKKTIITKSARDCHEFGNILHLSTNLVASIQRPDKHESFGNNMVDNLDLFSRSSAENKYDNGCAKLSpecifications
NP_008893 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
VEECPAEGKIPFWNFPEVCQVDEQIERQHQDDQDKCLLMQVGFSDKKTIITKSARDCHEFGNILHLSTNLVASIQRPDKHESFGNNMVDNLDLFSRSSAENKYDNGCAKL | |
RUO | |
ZNF182 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
7569 | |
ZNF21 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HHZ150/KOX14/MGC125383/MGC131713/ZNF21/Zfp182 | |
ZNF182 | |
Recombinant | |
wheat germ expression system |