missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ZNF69 Partial ORF (NP_068734.1, 1 a.a. - 81 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00007620-Q01.10ug

 View more versions of this product

Product Code. 16137775

  • 335.00 € / 10µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Sequence: MPCCSHRRCREDPGTSESQEMEEWALLDISQRKLYKEVMLETFRNLTSVGKSWKDQNIEYEYQNPRRNFRSLIEKKVNEIK
Specifications

Specifications

NP_068734.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
34.65kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MPCCSHRRCREDPGTSESQEMEEWALLDISQRKLYKEVMLETFRNLTSVGKSWKDQNIEYEYQNPRRNFRSLIEKKVNEIK
RUO
ZNF69
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
7620
ZNF69 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Cos5/MGC59928
ZNF69
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions