Learn More
HYDRANAL™ - Titrant 5 E, Reagent for volumetric two-component Karl Fischer titration (ethanol based), Honeywell™ Fluka™
Product Code. p-7072124 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
6 x 500 mL
Unit Size:
1 L
100mL
2.5 L
500mL
Pack of 6
Product Code. 15654370
Brand: Honeywell-Hydranal 34732100ML
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
Titrating agent for volumetric two-component Karl Fischer titration (ethanol-based)
Advantages of Hydranal two-component reagents:
  • High titration speed
  • Ideal accuracy for small amounts of water
  • High buffer capacity
  • Exact and stable titer
  • E-type reagents: reduced toxicity compared to methanol
  • Long shelf life (three years for titrants, five years for solvents)
Improved Composition
In two-component reagents the Karl Fisher reactants are separated into two solutions: the titrant and the solvent. Hydranal-Titrant contains iodine dissolved in an alcohol with a precisely defined concentration. Hydranal-Solvent is an alcoholic solution of sulfur dioxide and imidazole. The alcohol is either methanol for standard reagents or ethanol for E-type reagents.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material HYDRANAL™-Titrant 5 E
Density 0.89g/mL
Quantity 100 mL
Flash Point 12°C (53.6°F)
UN Number UN1170