Learn More
Thermo Scientific™ HyperSep™ Dispersive SPE Multipacks (QuEChERS)
Product Code. 12633085 Shop All Thermo Scientific Products
Click to view available options
Description:
1500 mg Sodium Chloride, 6000 mg Magnesium Sulfate, 50 pk
4000 mg Magnesium Sulfate, 1000 mg Sodium Acetate, 50 pk
4000 mg Magnesium Sulfate, 1000 mg Sodium Chloride, 500 mg Sodium Citrate dibasic sesquihydrate, 1000 mg sodium citrate tribasic, 50 pk
4000 mg Magnesium Sulfate, 1000 mg sodium chloride, 50 pk
6000 mg Magnesium Sulfate, 1500 mg NaCl, 1500 mg sodium citrate dihydrate, 750 mg disodium citrate sesquihydrate
6000 mg Magnesium Sulfate, 1500 mg Sodium Acetate, 50 pk
8000 mg Magnesium Sulfate, 2000 mg sodium chloride, 50 pk
8000 mg Magnesium Sulfate, 3500 mg NaCl, 50 pk
Unit Size:
Pack of 50
Product Code. 12633085
Brand: Thermo Scientific™ 60105335
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
Denne vare kan ikke returneres. Se returpolitik
Save time and achieve more accurate pesticide determinations using these pre-prepared dispersive SPE products for QuEChERS clean-up.
QuEChERS Dispersive SPE methods offer a convenient and effective approach for extraction and clean-up of pesticide residues in food and other complex matrices. The robust procedure offers a number of compelling advantages: high recoveries, accurate results, high sample throughput, low solvent and glassware usage, reduced labor and bench space and lower reagent costs.
Thermo Scientific™ HyperSep Dispersive SPE Clean-Up Products are available in a range of pre-prepared sorbent combinations, each containing the proper sorbents for optimum clean-up of analytes extracted using QuEChERS methods.
HyperSep Dispersive SPE Clean-Up Products contain PSA (primary/secondary amine) for the removal of organic acids and polar pigments among other compounds. Some products couple the PSA with endcapped C18 for the removal of most lipids and sterols, or graphitized carbon black for the removal of sterols and pigments such as chlorophyll. A variety of tube formats and bed weights are available to accommodate large and small sample sizes.
All HyperSep Dispersive SPE products are pre-prepared and optimized for the method at hand, so you will:
  • Save valuable preparation time
  • Achieve more consistent analytical results due to consistency in products and rigorous quality control
  • Get cleaner extracts and lower peak counts as a result of using highest quality, pre-prepared products
QuEChERS Technique
Developed by the USDA Eastern Regional Research Center in 2003, the QuEChERS (Quick, Easy, Cheap, Effective, Rugged and Safe) technique is increasingly becoming the technique of choice for extraction and clean-up of pesticide residues.
The QuEChERS method is a two-step process: extraction followed by clean-up. There are four variations of the QuEChERS method currently in use:
  • The original QuEChERS method uses sodium chloride to enhance extraction
  • Dispersive AOAC 2007.01 uses sodium acetate as a buffer, replacing sodium chloride
  • The Dual-Phase variation introduces PSA and GCB to remove high levels of chlorophyll and plant sterols in the final extract without the loss of planar pesticides (polar aromatics) using an acetone:toluene solvent mix (3:1)
  • European Version is similar to the AOAC method, except the extraction uses sodium chloride, sodium citrate dihydrate and disodium citrate sesquihydrate instead of sodium acetate
TRUSTED_SUSTAINABILITY
Tekniske data
Description 6000 mg Magnesium Sulfate, 1500 mg Sodium Acetate, 50 pk
For Use With (Application) QuEChERS Method
Product Line HyperSep
Sorbent Anhydrous Magnesium Sulfate, Sodium Acetate
Type QuEChERS Sample Prep Multipack
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.