Learn More
IBA1 Mouse anti-Human, Unconjugated, Clone: 1C6A10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66827-1-IG-150UL
Description
The IBA1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human IBA1. This antibody recognizes human antigen. The IBA1 antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
| IBA1 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Immunohistochemistry, Immunofluorescence | |
| 1C6A10 | |
| Unconjugated | |
| 150 μL | |
| 199 | |
| IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.