missing translation for 'onlineSavingsMsg'
Learn More

PeproTech IL-7 Polyclonal Antibody, PeproTech®, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  PeproTech 500-P27-50UG

 View more versions of this product

Product Code. 17895043

  • 219.00 € / 50µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

AA Sequence of recombinant protein: MDCDIEGKDG KQYESVLMVS IDQLLDSMKE IGSNCLNNEF NFFKRHICDA NKEGMFLFRA ARKLRQFLKM NSTGDFDLHL LKVSEGTTIL LNCTGQVKGR KPAALGEAQP TKSLEENKSL KEQKKLNDLC FLKRLLQEIK TCWNKILMGT KEH. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human IL-7. Anti-Human IL-7-specific antibody was purified by affinity chromatography employing an immobilized Human IL-7 matrix. Sandwich ELISA: To detect Human IL-7 by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human IL-7 (500-P27Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human IL-7. Western Blot: To detect Human IL-7 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. When used in conjunction with compatible secondary reagents, the detection limit for Recombinant Human IL-7 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.

Interleukin 7 (IL-7) is a hematopoietic cytokine that is constitutively produced by stromal cells from the bone marrow and thymus. IL-7 signals through the IL-7R to promote the differentiation of hematopoietic stem cells to lymphoid precursor cells, which give rise to T cells, B cells, and natural killer cells.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

IL-7
Polyclonal
Unconjugated
IL7r
A630026I06Rik; H-IL-7; hlb368; IL7; Il-7; ILN; Interleukin; interleukin 7; Interleukin7; interleukin-7; M-IL-7
Rabbit
Antigen affinity chromatography
RUO
3574, 3575
-20°C
Lyophilized
ELISA, Western Blot
0.1-1.0 mg/mL
PBS with no preservative
P13232, P16871
Il7, IL7r
E.coli-derived Recombinant Human IL-7.
50 μg
Primary
Human
Antibody
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.