missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ IL1F7 Recombinant Protein

Brand:  Abnova™ H00027178-P01.10ug

 View more versions of this product

Product Code. 16138642

  • 335.00 € / 10µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH20637
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
50.1
8
Glutathione Sepharose 4 Fast Flow
10 ug
MSFVGENSGVKMGSEDWEKDEPQCCLEDPAVSPLEPGPSLPAMNFVHTSPKVKNLNPKKFSIHDQDHKVLVLDSGNLIAVPDKNYIRPEIFFALASSLSSASAEKGSPILLGVSKGEFCLYCDKDKGQSHPSLQLKKEKLMKLAAQKESARRPFIFYRAQVGSWNMLESAAHPGWFICTSCNCNEPVGVTDKFENRKHIEFSFQPVCKAEMSPSEVSD
FIL1/FIL1(ZETA)/FIL1Z/IL-1F7/IL-1H4/IL-1RP1/IL1H4/IL1RP1
IL1F7
Wheat Germ (in vitro)
GST
Antibody Production, Protein Array, ELISA, Western Blot
27178
IL1F7 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
IL1F7
Human
Recombinant
Solution
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions