missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies ImmunoCult™-SF Macrophage Medium
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
Description
- ImmunoCult™-SF Macrophage Medium has been developed for the in vitro culture and differentiation of human monocytes into macrophages when the appropriate cytokines and stimuli are added
- The factors for differentiation and activation of macrophages have not been added to ImmunoCult™-SF Macrophage Medium
- This provides users the flexibility to prepare a medium that meets their requirements
- The medium is a specialized serum-free culture medium that can be used to differentiate human monocytes into M1 (classically activated) and M2a (alternatively activated) macrophages in a 6- or 8-day culture period
Specifications
Specifications
| Cell Type | Macrophages; Monocytes |
| For Use With (Application) | Cell Culture; Differentiation |
| Quantity | 250 mL |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction