Learn More
Fisherbrand™ Individual Stainless Steel Cylindrical Calibration Weights, Class E2
Product Code. 15585039 Shop All Fisherbrand Products
Click to view available options
Weight (Metric):
1 g
2 g
5 g
10 g
20 g
50 g
100 g
200 g
500 g
1 kg
2 kg
5 kg
10 kg
Unit Size:
1 unit
Product Code. 15585039
Please to purchase this item. Need a web account? Register with us today!
Calibrate weights of lower accuracy classes. Fisherbrand™ Class E2 Individual Stainless Steel Cylindrical Calibration Weights are intended for use in the verification or calibration of class F1 weights and for use with weighing instruments of special accuracy class I.
  • Class E2 stainless steel calibration weight
  • In compliance with OIML R111
Specifications
Classification E2
Material Stainless Steel
Weight (Metric) 20 g
Mass 20 g
Shape Cylindrical