Learn More
Smeg™ Injection Trolley for Laboratory Glassware Washers
Product Code. p-8011010
Click to view available options
Holds:
20 items
20 narrow-necked items, plus 20 pipettes
24 items
40 items
68 items
Includes:
20 spigots, drying system connection
40 U4140 spigots, without drying system connection
40 spigots, drying system connection
68 U4140 spigots and 1 UC6 spigot, drying system connection
Fittings for ULB 40 adaptor nozzles for pipettes
Without drying system connection
For Use With (Application):
For washing and drying narrow-necked Glassware up to 490mm height, flasks, round-bottom flasks and g
Internal injection washing of narrow-necked glassware with a maximum height of 225mm
Simultaneous washing of narrow-necked and other types of glassware
Suitable for washing and drying narrownecked glassware up to 19.3 inches / 490 mm in height.
Washing narrow-necked glassware of different sizes
Washing narrow-necked glassware up to 490mmH
Washing up to 68 glassware items
Unit Size:
Each
Product Code. 11915594
Brand: Smeg™ LM20DS
Please to purchase this item. Need a web account? Register with us today!
Use these trolleys with Smeg™ glassware washers. Smeg Injection Trolleys for Laboratory Glassware Washers are available in a variety of configurations to fit a wide range of applications.
  • For use with Smeg glassware washers
  • Available with one, two and mixed spraying/injection levels
  • Clean and sanitize a wide variety of labware
Specifications
Material Stainless Steel
Product Type Injection Trolley
Holds 20 items
Rack Type 20-position universal flask washing trolley in stainless steel with 20 spigots and drying system connection. Half the trolley space is free for mixed glassware suitable for a ½ basket accessory.
Style Injection trolley with one spraying/injection level
For Use With (Application) Suitable for washing and drying narrownecked glassware up to 19.3 inches / 490 mm in height.
For Use With (Equipment) Glassware washers
Includes 20 spigots, drying system connection
Quantity 1 Each