missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Inosine 5'-monophosphate, disodium salt hydrate, 97%

41.10 € - 379.89 €

Chemical Identifiers

CAS 352195-40-5
Molecular Formula C10H11N4Na2O8P
Molecular Weight (g/mol) 392.17
MDL Number MFCD00150372
InChI Key AANLCWYVVNBGEE-WCYUCLFNNA-L
Synonym inosine 5'-monophosphate disodium salt hydrate
PubChem CID 126963545
IUPAC Name disodium;[(2S,3S,4R,5R)-3,4-dihydroxy-5-(6-oxo-3H-purin-9-yl)oxolan-2-yl]methyl phosphate;hydrate
SMILES [Na+].[Na+].O[C@@H]1[C@@H](COP([O-])([O-])=O)O[C@H]([C@@H]1O)N1C=NC2=C1N=CNC2=O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10551281
SDS View Documents
Thermo Scientific Chemicals
226260050
5 g Glass bottle
41.10 €
5g
Please to purchase this item. Need a web account? Register with us today!
10236520
SDS View Documents
Thermo Scientific Chemicals
226260250
25 g Glass bottle
126.63 €
25g
Please to purchase this item. Need a web account? Register with us today!
10053523
SDS View Documents
Thermo Scientific Chemicals
226261000
100 g Glass bottle
379.89 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause damage to organs through prolonged or repeated exposure.
Suspected of damaging fertility.

GHS P Statement:
Use personal protective equipment as required.
Do not breathe dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

Product Identifier
  • Inosine 5'-monophosphate, disodium salt hydrate

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Inosine 5'-monophosphate, disodium salt hydrate, 97%

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.