Learn More
B Braun™ FEP Safety IV Catheters, Introcan
Product Code. 10174713
Click to view available options
Needle Gauge:
18 G
22 G
24 G
missing translation for 'unitSize'
Pack of 50
Product Code. 10174713
missing translation for 'mfr': B Braun™ 4252098B
Please to purchase this item. Need a web account? Register with us today!
Use B Braun™ FEP Safety IV Catheters, Introcan™. From insertion to catheter deployment to needle removal, protection is provided by an automatically activated system that cannot be bypassed.
  • Color-coded labels for easy gauge size identification
  • Back cut bevel
  • FEP for firmer construction for arterial access and special procedures
  • DEHP- and latex-free
Specifications
Material FEP
Needle Gauge 22 G
Product Type Introcan