Learn More
Invitrogen™ Flp-In™ Core System
Click to view available options
Quantity:
1 kit
Unit Size:
Each
Description
Includes
The Flp-In Core Kit includes 20μg each of pFRT/lacZeo, pOG44, pcDNA5/FRT, and pcDNA5/FRT/CAT (expression control), and 2μg each of forward and reverse sequencing primers.
- For introduction of Flp Recombination Target (FRT) site into genome of mammalian cell line of choice
- Expression vector containing gene of interest is then integrated into genome via Flp recombinase-mediated DNA recombination at FRT site
Constitutive Expression, Mammalian Expression, Protein Expression, Proteins, Expression, Isolation and Analysis, Targeted Integration
Specifications
Specifications
| Constitutive or Inducible System | Constitutive |
| Delivery Type | Transfection |
| Promoter | CMV |
| Product Type | Flp-In Core System |
| Selection Agent (Eukaryotic) | Zeocin™ |
| Content And Storage | The Flp-In™ Core Kit includes 20 μg each of pFRT⁄lacZeo, pOG44, pcDNA™5⁄FRT, and pcDNA™5⁄FRT⁄CAT (expression control), and 2 μg each of forward and reverse sequencing primers. Store at -20C. |
| Protein Tag | Untagged |
| Cloning Method | Restriction Enzyme/MCS |
| Quantity | 1 kit |
| Vector | pFRT |
| Show More |