missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ pEF5/FRT/V5-DEST™ Gateway™ Vector

Product Code. 11550856
Click to view available options
Quantity:
6 μg
Unit Size:
6µg
This item is not returnable. View return policy

For Research Use Only. All usage must comply with product instructions.

Product Code. 11550856

Brand: Invitrogen™ V602020

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For Research Use Only. All usage must comply with product instructions.

Includes

All destination vectors are provided lyophilized and supercoiled.

Requires

Additional materials required, available separately: Gateway entry clone, Gateway LR Clonase enzyme mix, and reaction buffer.

Destination vectors have attR sites for recombination with any attL-flanked fragment

Cloning, Constitutive Expression, Gateway Cloning, Mammalian Expression, Protein Expression, Proteins, Expression, Isolation and Analysis, Targeted Integration

TRUSTED_SUSTAINABILITY

Specifications

Constitutive or Inducible System Constitutive
Delivery Type Transfection
Promoter EF-1α
Product Type Gateway System Expression Vector
Selection Agent (Eukaryotic) Hygromycin
Content And Storage All destination vectors are provided lyophilized and supercoiled.
Protein Tag V5 Epitope Tag
Cloning Method Gateway
Quantity 6 μg
Vector pDEST
Product Line Flp-In, Gateway
For Use With (Application) Targeted Integration
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.