Learn More
Invitrogen™ SureCast™ Resolving Buffer
Product Code. p-6779027
Click to view available options
Quantity:
2 x 500 mL
5 x 500 mL
Unit Size:
Pack of 2
Pack of 5
Product Code. 15527935
Brand: Invitrogen™ HC2212
Please to purchase this item. Need a web account? Register with us today!
These components of the SureCast Handcast System can be used to create polyacrylamide gels using SureCast Handcast Reagents as well as any other popular polyacrylamide gel casting reagents.
  • Pour polyacrylamide gels easily and confidently
  • For use with the SureCast vertical gel system
  • SureCast gels are designed for use in the Mini Gel Tank
Recommended Storage
Room Temperature
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 2 x 500 mL (1 L total) SureCast Resolving Buffer packs.
Store at room temperature.
Product Type Resolving Buffer (1 L)
Product Line SureCast
Quantity 2 x 500 mL
Shipping Condition Room Temperature
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado