Learn More
Cytiva IPGbox
Tools for enhancing reswelling of precast Immobiline DryStrip gels (IPG strips)
Brand:  Cytiva 28-9334-65
Product Code. 10320785
  • 459.00 € / Each
Estimated Shipment: 04-03-2025
to see stock.
Explore more special offers
Add to basket
Includes: IPGbox: 1 IPGbox, 1 IPGbox Kit (10 Reswell trays, 1 IPGbox insert, instructions)
Description
Description
  • Oil free rehydratation of up to 12 Immobiline DryStrip gels or other IPG strips independently and simultaneously
  • Lid protects IPG strips from dust and contamination
  • Load samples for IEF during rehydration by including them in the rehydration bufferor load sample using sample cups
  • IPGbox used together with IPGbox Kit (Cat. No 45002627) to rehydrate 7 to 24cm Immobiline DryStrip gels
  • Includes reswelling tray made of hydrophobic material to prevent sticking of rehydration solution
  • IPGbox Insert located within lid of IPGbox to maintain optimal conditions for rehydration
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IPGBox
1 IPGbox and 1 IPGbox kit
Up to 12 Immobiline dry strip gel
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cytiva IPGbox > IPGbox; Includes: 1 IPGbox, 1 IPGbox Kit (10 Reswell trays, 1 IPGbox insert, instructions)