Learn More
Invitrogen™ iPrep™ Card: Viral DNA/RNA
Product Code. 10317654
Click to view available options
Quantity:
1 card
Unit Size:
Each
For Research Use Only. All usage must comply with product instructions.
Product Code. 10317654
missing translation for 'mfr': Invitrogen™ IS10016
Please to purchase this item. Need a web account? Register with us today!
For Research Use Only. All usage must comply with product instructions.
iPrep™ Card: Viral DNA/RNA
The iPrep™ Viral DNA/RNA protocol Card is for use with the iPrep™ PureLink™ Virus Kit. The iPrep™ Protocol Card is a flash, 512 KB memory card. Each iPrep™ Protocol card is pre-programmed with the purification protocol that directs the volume of reagents used and incubation time. The following iPrep™ Protocol Cards are available from Invitrogen:
  • iPrep™ Forensic Card for use with the iPrep™ ChargeSwitch™ Forensic Kit and the iPRep™ ChageSwitch™ gDNA Buccal kit
  • iPrep™ Tissue Card for use with the iPrep™ ChargeSwitch™ Tissue Kit
  • iPrep™ Blood Card for use with the iPrep™ PureLink™ Blood kit
  • iPrep™ Viral DNA/RNA Card for use with the iPrep™ PureLink™ Virus Kit
  • TRUSTED_SUSTAINABILITY
    Specifications
    Format Card
    Product Line iPrep
    Quantity 1 card