Learn More
Masterflex™ IQ/OQ Masterflex Pumps
Click to view available options
:
Assay Service Type: IQ/OQ
Description
Documented collection of activities necessary to:
- Estabilish if an instrument is delivered as designed and specified
- Determine if it is properly installed in the selected environment
- Ensuring that it will function according to its operational specifications in the selected environment
Specifications
Specifications
| For Use With (Equipment) | Masterflex™ Pumps |
| Assay Service Type | IQ/OQ |