missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl cyanoacetate, 97%, Thermo Scientific Chemicals

30.55 € - 69.55 €

Chemical Identifiers

CAS 13361-30-3
Molecular Formula C6H9NO2
Molecular Weight (g/mol) 127.143
MDL Number MFCD00019842
InChI Key BESQLCCRQYTQQI-UHFFFAOYSA-N
Synonym isopropyl cyanoacetate, isopropyl 2-cyanoacetate, cyanoacetic acid isopropyl ester, acetic acid, cyano-, isopropyl ester, unii-0v3mgi9xma, 0v3mgi9xma, acetic acid, cyano-, 1-methylethyl ester, propan-2-yl cyanoacetate, prop-2-yl cyanoacetate, acmc-1bsph
PubChem CID 25917
IUPAC Name propan-2-yl 2-cyanoacetate
SMILES CC(C)OC(=O)CC#N
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11403864
SDS View Documents
Thermo Scientific Alfa Aesar
A11887.14
25 g
30.55 €
25g
Please to purchase this item. Need a web account? Register with us today!
11413864
SDS View Documents
Thermo Scientific Alfa Aesar
A11887.22
100 g
69.55 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Isopropyl cyanoacetate was used in the synthesis of 3-substituted coumarins by undergoing KnÖevenagel reactions with aldehydes and ketones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isopropyl cyanoacetate was used in the synthesis of 3-substituted coumarins by undergoing Knöevenagel reactions with aldehydes and ketones.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry conditions in well sealed container. Store away from oxidizing agent.
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335

missing translation for 'einecsNumber' : 236-421-3

missing translation for 'rtecsNumber' : AG4270000

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Isopropyl cyanoacetate, 97%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.