Learn More
GPE Scientific™ J Young Sample Flask, Cylindrical with Fine Thread Tap and B14 Socket
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
missing translation for 'unitSize'
Each
Description
- Solidly constructed for use with vacuum manifolds
- Designed for use with Schlenk lines
- Includes fine thread tap and B14 socket
Specifications
Specifications
| Capacity (Metric) | 250 mL |
| For Use With (Equipment) | Vacuum manifolds and Schlenk lines |
| Includes | Fine thread tap and B14 socket |
| Product Type | Sample Flask |
| Socket Joint | Size: 14 |