Learn More
GPE Scientific™ J Young Schlenk Flask, Round Bottom with Glass Key and Socket
GPE Scientific™ J Young Schlenk Flask, Round Bottom with Glass Key and Socket is specially designed for use with Schlenk lines and vacuum manifolds, and is available in different capacities.
Brand: GPE Scientific™ SRG/50
This item is not returnable.
View return policy
Description
- Includes glass key and socket
- Available in various capacities
- Available with different joint sizes
Specifications
50 mL | |
Schlenk Flask | |
Round |
Glass key and socket | |
Joint size: 19 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
GPE Scientific™ J Young Schlenk Flask, Round Bottom with Glass Key and Socket > 50mL
Spot an opportunity for improvement?Share a Content Correction