missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JAM-A Antibody (RM0116-1M21), Alexa Fluor™ 647, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-22494AF647
This item is not returnable.
View return policy
Description
JAM-A Monoclonal specifically detects JAM-A in Mouse samples. It is validated for Western Blot.
Specifications
| JAM-A | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| F11R | |
| Mouse recombinant protein of the extracellular domain of JAM-A | |
| 0.1 mL | |
| 50848 | |
| Mouse | |
| Purified |
| Western Blot | |
| RM0116-1M21 | |
| Western Blot | |
| CD321 antigen, F11 receptor, JAM-1, JAM1CD321, JAMA, JAM-A, JCAMJAM, Junctional adhesion molecule 1Platelet F11 receptor, junctional adhesion molecule A, PAM-1KAT, Platelet adhesion molecule 1 | |
| Rat | |
| Protein A or G purified | |
| Primary | |
| This was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant protein of extracellular domain of JAM-A. | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction